Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tigd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198634.1 | NP_941036.1 | 2 | 373 | ||
GenBank mRNA | BC053500.2 | - | 2 | 412 | 30007514 | |
BC055784.1 | - | 2 | 373 | 6404652 |