Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198680.2 | NP_941373.1 | 8 | 1514 | ||
GenBank mRNA | AK003220.1 | - | 8 | 1515 | ||
BC099584.1 | - | 8 | 1511 | 30291007 |