Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005780.2 | NP_005771.1 | 2 | 767 | ||
XM_011534861.1 | XP_011533163.1 | 2 | 570 | |||
GenBank mRNA | AF098807.1 | - | 2 | 612 | ||
AK313036.1 | - | 2 | 719 | |||
BC017824.1 | - | 2 | 615 | 4696562 |