Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013854.1 | NP_038882.1 | 4 | 278 | ||
GenBank mRNA | AK077741.1 | - | 4 | 290 | ||
BC032250.1 | - | 4 | 271 | |||
BC046965.1 | - | 4 | 252 | 6485365 | ||
BC063094.1 | - | 4 | 278 | 6516514 |