Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024454.1 | NP_077774.1 | 6 | 713 | ||
GenBank mRNA | AB232615.1 | - | 6 | 468 | ||
AK035980.1 | - | 5 | 282 | |||
AK172891.1 | - | 6 | 713 | |||
BC055042.1 | - | 6 | 713 | 6490069 |