Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252468.1 | NP_001239397.1 | 3 | 467 | ||
NM_001252469.1 | NP_001239398.1 | 3 | 273 | |||
NM_057171.2 | NP_476512.1 | 3 | 467 | |||
GenBank mRNA | AK148796.1 | - | 3 | 387 | ||
AK164266.1 | - | 3 | 244 | |||
AK170134.1 | - | 3 | 287 | |||
BB857758.1 | - | 3 | 208 | |||
BC037051.1 | - | 3 | 291 | |||
BC048137.1 | - | 3 | 316 |