Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139200.4 | NP_631939.1 | 5 | 522 | ||
GenBank mRNA | AF192525.1 | - | 5 | 442 | ||
AK089364.1 | - | 5 | 463 | |||
AK152784.1 | - | 5 | 504 | |||
AK157859.1 | - | 5 | 523 | |||
AK164790.1 | - | 6 | 528 | |||
AK166254.1 | - | 5 | 523 | |||
AK168343.1 | - | 5 | 522 | |||
AK169575.1 | - | 5 | 508 | |||
AK170544.1 | - | 5 | 523 | |||
AK170796.1 | - | 5 | 523 | |||
AK171467.1 | - | 5 | 523 | |||
AK172071.1 | - | 5 | 579 | |||
AK172380.1 | - | 5 | 523 | |||
BC007144.1 | - | 5 | 511 | 3487179 |