Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290634.1 | NP_001277563.1 | 10 | 1938 | ||
NM_139297.6 | NP_647458.1 | 10 | 2128 | |||
GenBank mRNA | AF424698.1 | - | 10 | 1976 | ||
AK033445.1 | - | 10 | 1975 | |||
AK177417.1 | - | 1 | 302 | |||
BC023779.1 | - | 3 | 577 | |||
BC025585.1 | - | 10 | 1883 | 5324928 | ||
BC026626.1 | - | 9 | 1665 | |||
BC061208.1 | - | 10 | 2015 | 30250244 |