Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144500.3 | NP_653083.2 | 14 | 1913 | ||
GenBank mRNA | AK047279.1 | - | 1 | 885 | ||
AK048537.1 | - | 14 | 1833 | |||
AK145292.1 | - | 14 | 1946 | |||
AK161695.1 | - | 14 | 1856 | |||
AK170306.1 | - | 14 | 1897 | |||
AK170669.1 | - | 14 | 1914 | |||
AK183662.1 | - | 1 | 47 | |||
AK220257.1 | - | 8 | 1232 | |||
BC026804.1 | - | 14 | 1920 | 4159217 |