Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144551.5 | NP_653134.2 | 2 | 1686 | ||
GenBank mRNA | AF358867.1 | - | 2 | 551 | ||
AK044747.1 | - | 2 | 1644 | |||
AK080064.1 | - | 2 | 889 | |||
AK082329.1 | - | 2 | 898 | |||
AK144192.1 | - | 2 | 1751 | |||
BC027159.1 | - | 1 | 143 | |||
BC034338.1 | - | 2 | 720 | 5043179 | ||
BC037387.1 | - | 2 | 1630 | 5363174 |