Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144551.5 | NP_653134.2 | 3 | 4006 | ||
GenBank mRNA | AK080064.1 | - | 3 | 3210 | ||
AK082329.1 | - | 3 | 3217 | |||
AK144192.1 | - | 3 | 4070 | |||
BC027159.1 | - | 2 | 2462 | |||
BC034338.1 | - | 3 | 3039 | 5043179 | ||
BC037387.1 | - | 3 | 3950 | 5363174 |