Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146180.1 | NP_001139652.1 | 5, 6 | 882 | ||
NM_144800.2 | NP_659049.2 | 5, 6 | 882 | |||
GenBank mRNA | AK030533.1 | - | 3, 4 | 276 | ||
AK129139.1 | - | 5, 6 | 879 | |||
AY214918.1 | - | 5, 6 | 383 | |||
BC024131.1 | - | 5, 6 | 847 | 5101888 |