Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144800.2 | NP_659049.2 | 11, 12 | 1543 | ||
GenBank mRNA | AK030533.1 | - | 9, 10 | 937 | ||
AY214918.1 | - | 11, 12 | 1044 | |||
BC023682.1 | - | 5, 6 | 649 | |||
BC024131.1 | - | 11, 12 | 1508 | 5101888 |