Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144804.1 | NP_659053.1 | 2 | 226 | ||
GenBank mRNA | AK134830.1 | - | 2 | 246 | ||
AK144066.1 | - | 2 | 239 | |||
AK162086.1 | - | 2 | 249 | |||
AK162218.1 | - | 2 | 249 | |||
AK165504.1 | - | 2 | 239 | |||
BC013499.1 | - | 2 | 226 | 4222591 |