Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164242.1 | NP_001157714.1 | 4, 5 | 405 | ||
NM_001164243.1 | NP_001157715.1 | 4, 5 | 448 | |||
NM_001164244.1 | NP_001157716.1 | 3, 4 | 352 | |||
NM_144806.2 | NP_659055.1 | 5, 6 | 501 | |||
GenBank mRNA | AK038680.1 | - | 5, 6 | 467 | ||
AK076811.1 | - | 5, 6 | 455 | |||
AK077330.1 | - | 5, 6 | 454 | |||
BC023178.1 | - | 5, 6 | 486 | 4947226 | ||
BC051438.1 | - | 4, 5 | 372 | 6311509 | ||
BC106185.1 | - | 4, 5 | 334 | 6512229 |