Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318875.1 | NP_001305804.1 | 4 | 509 | ||
NM_005803.3 | NP_005794.1 | 4 | 509 | |||
XM_005248780.3 | XP_005248837.1 | 4 | 242 | |||
XM_017010157.1 | XP_016865646.1 | 4 | 163 | |||
GenBank mRNA | AF085357.1 | - | 4 | 298 | ||
AF089750.2 | - | 4 | 334 | |||
AF117234.1 | - | 4 | 335 | |||
AK000816.1 | - | 4 | 345 | |||
AK222530.1 | - | 4 | 345 | |||
AK301291.1 | - | 4 | 343 | |||
BC001146.1 | - | 3 | 192 | 3534771 | ||
BT007077.1 | - | 3 | 170 | |||
CR456946.1 | - | 3 | 170 |