Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135151.1 | NP_001128623.1 | 10 | 3575 | ||
NM_001135152.1 | NP_001128624.1 | 10 | 3541 | |||
NM_144808.4 | NP_659057.2 | 10 | 3575 | |||
GenBank mRNA | AB177995.1 | - | 11 | 2292 | ||
AK122197.1 | - | 7 | 3038 | |||
BC030883.1 | - | 1 | 1522 |