Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144813.1 | NP_659062.1 | 2 | 442 | ||
GenBank mRNA | BC016094.1 | - | 2 | 442 | 4504496 |