Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005804.3 | NP_005795.2 | 2, 3 | 333 | ||
NR_046366.2 | 2, 3 | 333 | ||||
XM_006722606.3 | XP_006722669.1 | 2, 3 | 273 | |||
XM_011527620.1 | XP_011525922.1 | 2, 3 | 352 | |||
XM_011527621.2 | XP_011525923.1 | 1, 2 | 219 | |||
GenBank mRNA | AB254849.1 | - | 1, 2 | 215 | ||
AK301847.1 | - | 3, 4 | 416 | |||
BC001009.2 | - | 2, 3 | 270 | 2820926 | ||
BC032128.2 | - | 2, 3 | 279 | 5017293 | ||
DA432925.1 | - | 2, 3 | 334 | |||
DB032777.1 | - | 3, 4 | 426 | |||
U90426.1 | - | 2, 3 | 310 |