Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncaph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144818.3 | NP_659067.2 | 12, 13 | 1660 | ||
GenBank mRNA | AK028919.1 | - | 12, 13 | 1645 | ||
AK042706.1 | - | 12, 13 | 1577 | |||
AK080433.1 | - | 12, 13 | 1649 | |||
AK129047.1 | - | 9, 10 | 1213 | |||
BC021499.1 | - | 12, 13 | 1634 | 5346298 | ||
BC025060.1 | - | 12, 13 | 1585 | 3673890 |