Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144835.4 | NP_659084.3 | 7 | 991 | ||
GenBank mRNA | AK044969.1 | - | 7 | 987 | ||
AK087792.1 | - | 7 | 996 | |||
AK143958.1 | - | 7 | 969 | |||
AK163223.1 | - | 7 | 955 | |||
AK165368.1 | - | 7 | 933 | |||
AK165890.1 | - | 7 | 994 | |||
AK172508.1 | - | 7 | 988 | |||
BC024471.1 | - | 4 | 442 |