Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144846.5 | NP_659095.1 | 4, 5 | 494 | ||
GenBank mRNA | AK028062.1 | - | 4, 5 | 495 | ||
AK075613.1 | - | 5, 6 | 439 | |||
BC011343.1 | - | 4, 5 | 312 | 4012217 |