Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144849.1 | NP_659098.1 | 8 | 1567 | ||
GenBank mRNA | AK050440.1 | - | 7 | 2282 | ||
AK133785.1 | - | 8 | 1591 | |||
AK143625.1 | - | 8 | 1595 | |||
BC014743.1 | - | 8 | 1567 | 4503319 | ||
DQ884878.1 | - | 8 | 1595 |