Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144853.3 | NP_659102.1 | 4 | 630 | ||
GenBank mRNA | AF442733.1 | - | 4 | 365 | ||
AK031840.1 | - | 4 | 654 | |||
AY061854.1 | - | 4 | 652 | |||
BC099957.1 | - | 4 | 630 | 6431603 | ||
BC125594.1 | - | 4 | 424 | 40130109 | ||
BC125596.1 | - | 4 | 424 | 40130111 |