Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025309.1 | NP_001020480.1 | 4 | 889 | ||
NM_144859.2 | NP_659108.1 | 4 | 889 | |||
GenBank mRNA | AF493070.1 | - | 4 | 946 | ||
AK122282.1 | - | 4 | 848 | |||
AK144586.1 | - | 4 | 876 | |||
AK164025.1 | - | 4 | 894 | |||
AK168371.1 | - | 4 | 871 | |||
BC017130.1 | - | 3 | 815 | 4506259 |