Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110015.1 | NP_001103485.1 | 12 | 1442 | ||
NM_001110016.1 | NP_001103486.1 | 12 | 1442 | |||
NM_144863.4 | NP_659112.2 | 12 | 1442 | |||
GenBank mRNA | AK041763.1 | - | 12 | 1362 | ||
AK045396.1 | - | 12 | 1415 | |||
AK077545.1 | - | 12 | 1371 | |||
AK155965.1 | - | 12 | 1340 | |||
AK156822.1 | - | 1443 | ||||
AK171683.1 | - | 12 | 1380 | |||
AK172054.1 | - | 12 | 1367 | |||
AK172377.1 | - | 12 | 1372 | |||
AK186642.1 | - | 1 | 37 | |||
AK214226.1 | - | 1 | 37 | |||
BC022132.1 | - | 12 | 1414 | |||
BC025809.1 | - | 12 | 1356 | |||
BC094453.1 | - | 12 | 1393 | 5068941 |