Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204914.1 | NP_001191843.1 | 8 | 1899 | ||
NM_144865.2 | NP_659114.2 | 8 | 1905 | |||
GenBank mRNA | AK147333.1 | - | 2 | 6695 | ||
AK158694.1 | - | 3 | 1479 | |||
BB709634.1 | - | 1 | 397 | |||
BC020184.1 | - | 8 | 1836 | 4505679 |