Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039482.1 | NP_001034571.1 | 6 | 1033 | ||
GenBank mRNA | AK032131.1 | - | 4 | 520 | ||
AK041813.1 | - | 1 | 1286 | |||
AK053102.1 | - | 6 | 983 | |||
AK054169.1 | - | 6 | 1001 | |||
AK220547.1 | - | 6 | 1012 | |||
BC019571.1 | - | 6 | 1005 | 4235202 | ||
BC060183.1 | - | 6 | 962 |