Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039482.1 | NP_001034571.1 | 12 | 2452 | ||
GenBank mRNA | AK032131.1 | - | 10 | 1942 | ||
AK046442.1 | - | 3 | 752 | |||
AK050964.1 | - | 1 | 181 | |||
AK053102.1 | - | 12 | 2404 | |||
BC029801.1 | - | 6 | 1175 | |||
BC060183.1 | - | 11 | 3126 |