Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144880.4 | NP_659129.2 | 7 | 1440 | ||
GenBank mRNA | AK145397.1 | - | 8 | 826 | ||
BC059026.1 | - | 7 | 1440 | 6810754 |