Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mavs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206382.1 | NP_001193311.1 | 5 | 615 | ||
NM_001206383.1 | NP_001193312.1 | 3 | 349 | |||
NM_001206385.1 | NP_001193314.1 | 4 | 524 | |||
NM_144888.2 | NP_659137.1 | 4 | 752 | |||
GenBank mRNA | AK028421.1 | - | 4 | 523 | ||
AK085038.1 | - | 4 | 711 | |||
AK153748.1 | - | 5 | 617 | |||
BC020006.1 | - | 4 | 484 | 4190175 | ||
BC025825.1 | - | 4 | 677 | 5100990 | ||
BC031352.1 | - | 4 | 661 | 5123637 | ||
BC037391.1 | - | 3 | 463 | 5361522 | ||
DQ167127.1 | - | 3 | 401 | |||
DQ174271.1 | - | 3 | 401 |