Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mavs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206382.1 | NP_001193311.1 | 8 | 1363 | ||
NM_001206383.1 | NP_001193312.1 | 6 | 1097 | |||
NM_001206385.1 | NP_001193314.1 | 7 | 1272 | |||
NM_144888.2 | NP_659137.1 | 7 | 1500 | |||
GenBank mRNA | AK085038.1 | - | 7 | 1461 | ||
AK153748.1 | - | 8 | 1365 | |||
AK181725.1 | - | 1 | 56 | |||
AK193001.1 | - | 1 | 56 | |||
AX775773.1 | - | 7 | 1249 | |||
BC020006.1 | - | 7 | 1232 | 4190175 | ||
BC025825.1 | - | 7 | 1425 | 5100990 | ||
BC031352.1 | - | 7 | 1409 | 5123637 | ||
BC037391.1 | - | 6 | 1211 | 5361522 | ||
DQ174271.1 | - | 6 | 1149 |