Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252573.1 | NP_001239502.1 | 2 | 439 | ||
NM_001252574.1 | NP_001239503.1 | 2 | 374 | |||
NM_001252575.1 | NP_001239504.1 | 2 | 393 | |||
NM_144893.2 | NP_659142.1 | 2 | 393 | |||
GenBank mRNA | AK053180.1 | - | 2 | 380 | ||
AK155539.1 | - | 2 | 408 | |||
AK158331.1 | - | 2 | 326 | |||
AK160899.1 | - | 2 | 379 | |||
AK170715.1 | - | 2 | 344 | |||
BB849961.1 | - | 2 | 326 | |||
BC018327.1 | - | 3 | 259 | 4166249 | ||
BC094025.1 | - | 2 | 372 | 4488690 |