Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252573.1 | NP_001239502.1 | 10 | 1595 | ||
NM_001252574.1 | NP_001239503.1 | 10 | 1530 | |||
NM_001252575.1 | NP_001239504.1 | 10 | 1478 | |||
NM_144893.2 | NP_659142.1 | 10 | 1549 | |||
GenBank mRNA | AK053180.1 | - | 10 | 1465 | ||
AK155539.1 | - | 10 | 1564 | |||
AK160899.1 | - | 10 | 1535 | |||
AK170715.1 | - | 10 | 1500 | |||
BB772752.1 | - | 1 | 88 | |||
BC018327.1 | - | 11 | 1415 | 4166249 | ||
BC064092.1 | - | 2 | 682 | |||
BC094025.1 | - | 10 | 1528 | 4488690 |