Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144987.1 | NP_001138459.1 | 2 | 460 | ||
NM_001144988.1 | NP_001138460.1 | 2 | 244 | |||
NM_144895.2 | NP_659144.1 | 2 | 244 | |||
GenBank mRNA | AK035081.1 | - | 1 | 103 | ||
AK087388.1 | - | 2 | 211 | |||
AK089946.1 | - | 2 | 244 | |||
AK129171.1 | - | 2 | 223 | |||
AK147760.1 | - | 2 | 461 | |||
AK159951.1 | - | 2 | 245 | |||
BC022921.1 | - | 2 | 212 | 4220506 |