Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161854.1 | NP_001155326.1 | 7 | 1115 | ||
NM_144901.3 | NP_659150.1 | 8 | 1208 | |||
GenBank mRNA | AK122393.1 | - | 7 | 1025 | ||
AK145351.1 | - | 7 | 1220 | |||
AK145428.1 | - | 7 | 1037 | |||
AK145932.1 | - | 7 | 1032 | |||
AK146586.1 | - | 7 | 1054 | |||
AK146706.1 | - | 8 | 1140 | |||
AK147012.1 | - | 7 | 1054 | |||
AK150882.1 | - | 7 | 1032 | |||
AK151638.1 | - | 7 | 1051 | |||
AK151723.1 | - | 7 | 1055 | |||
AK151970.1 | - | 7 | 1055 | |||
AK152531.1 | - | 7 | 1051 | |||
AK152777.1 | - | 3 | 262 | |||
AK153196.1 | - | 7 | 1032 | |||
AK153263.1 | - | 3 | 262 | |||
AK154068.1 | - | 7 | 1040 | |||
AK155751.1 | - | 8 | 1133 | |||
AK159258.1 | - | 7 | 1048 | |||
AK159863.1 | - | 7 | 1048 | |||
AK168556.1 | - | 6 | 849 | |||
BC016898.1 | - | 7 | 1052 | 4224466 | ||
BC024826.1 | - | 4 | 335 | |||
BC029058.1 | - | 2 | 124 | 4947555 | ||
BC029688.1 | - | 7 | 1051 | 5367783 | ||
BC031404.1 | - | 7 | 1023 | 4953772 | ||
BC036720.1 | - | 4 | 341 | |||
BC062097.1 | - | 8 | 1123 | 6833505 |