Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sesn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144907.1 | NP_659156.1 | 4 | 785 | ||
GenBank mRNA | AK143788.1 | - | 4 | 829 | ||
AK170228.1 | - | 4 | 798 | |||
AK170547.1 | - | 829 | ||||
AK196889.1 | - | 1 | 14 | |||
BC005672.1 | - | 4 | 785 | 3153129 |