Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sesn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144907.1 | NP_659156.1 | 6 | 1089 | ||
GenBank mRNA | AK143788.1 | - | 6 | 1133 | ||
AK170228.1 | - | 6 | 1102 | |||
AK170547.1 | - | 1133 | ||||
AK177455.1 | - | 1 | 16 | |||
AK179479.1 | - | 1 | 16 | |||
BC005672.1 | - | 6 | 1089 | 3153129 |