Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145138.2 | NP_660120.2 | 17, 18 | 2308 | ||
GenBank mRNA | AJ489828.1 | - | 17, 18 | 2283 | ||
AK173323.1 | - | 17, 18 | 2292 | |||
BC117971.2 | - | 17, 18 | 2249 | 40098375 | ||
BC117972.2 | - | 17, 18 | 2248 | 40098378 |