Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145148.2 | NP_660130.2 | 3, 4 | 664 | ||
GenBank mRNA | AB042027.1 | - | 3, 4 | 587 | ||
AF327856.1 | - | 2, 3 | 287 | |||
AF327857.1 | - | 1, 2 | 74 | |||
AK051779.1 | - | 3, 4 | 536 | |||
AK129262.1 | - | 2, 3 | 139 | |||
AK132280.1 | - | 3, 4 | 528 | |||
AK143978.1 | - | 2, 3 | 286 | |||
BC052390.1 | - | 3, 4 | 533 | 5718942 |