Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145148.2 | NP_660130.2 | 8, 9 | 1003 | ||
GenBank mRNA | AB042027.1 | - | 8, 9 | 926 | ||
AF327856.1 | - | 7, 8 | 626 | |||
AF327857.1 | - | 6, 7 | 413 | |||
AK129262.1 | - | 7, 8 | 478 | |||
AK132280.1 | - | 8, 9 | 867 | |||
AK143978.1 | - | 7, 8 | 625 | |||
BC052390.1 | - | 8, 9 | 872 | 5718942 |