Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145148.2 | NP_660130.2 | 11 | 1197 | ||
GenBank mRNA | AB042027.1 | - | 11 | 1120 | ||
AF327856.1 | - | 10 | 820 | |||
AF327857.1 | - | 9 | 607 | |||
AK129262.1 | - | 10 | 672 | |||
AK132280.1 | - | 11 | 1061 | |||
AK143978.1 | - | 10 | 819 | |||
BC052390.1 | - | 11 | 1066 | 5718942 |