Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145209.3 | NP_660210.1 | 7 | 1204 | ||
GenBank mRNA | AB067533.1 | - | 6 | 1129 | ||
AF426289.1 | - | 6 | 1135 | |||
AK078690.1 | - | 7 | 1205 | |||
AK089111.1 | - | 6 | 1176 | |||
AK149824.1 | - | 6 | 1128 | |||
AK150863.1 | - | 6 | 1258 | |||
AK165578.1 | - | 6 | 1241 | |||
AK172183.1 | - | 2 | 1628 | |||
AY057107.1 | - | 6 | 1117 | |||
AY089728.1 | - | 6 | 1129 | |||
BC032152.1 | - | 6 | 1141 | 5068699 |