Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145218.3 | NP_660253.2 | 4 | 1578 | ||
GenBank mRNA | AB175684.1 | - | 4 | 656 | ||
AF467979.1 | - | 4 | 859 | |||
AK035817.1 | - | 4 | 1332 | |||
AK048758.1 | - | 4 | 1323 | |||
AK051281.1 | - | 4 | 1978 | |||
AK081848.1 | - | 4 | 972 | |||
AK087726.1 | - | 2148 | ||||
BC052469.1 | - | 4 | 974 | 6810287 | ||
BC069177.1 | - | 4 | 1578 | |||
BC158110.1 | - | 4 | 1605 | 9088184 |