Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lactb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145381.2 | NP_663356.1 | 1, 2 | 186 | ||
GenBank mRNA | AK088932.1 | - | 1, 2 | 199 | ||
AK090091.1 | - | 1, 2 | 215 | |||
AK143912.1 | - | 1, 2 | 165 | |||
BC004045.1 | - | 1, 2 | 186 | 3496398 |