Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Athl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145387.4 | NP_663362.2 | 2 | 225 | ||
GenBank mRNA | AK077662.1 | - | 2 | 226 | ||
AK085600.1 | - | 2 | 241 | |||
AK220240.1 | - | 2 | 185 | |||
BC056953.1 | - | 2 | 187 | 5717810 |