Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnpo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122843.1 | NP_001116315.1 | 9 | 937 | ||
NM_145390.4 | NP_663365.3 | 9 | 919 | |||
GenBank mRNA | AK142270.1 | - | 8 | 1474 | ||
AK154985.1 | - | 9 | 921 | |||
AK155760.1 | - | 9 | 921 | |||
BC003275.1 | - | 9 | 873 | 3499838 | ||
BQ771085.1 | - | 2 | 109 |