Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tapbpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145391.2 | NP_663366.2 | 4 | 1160 | ||
GenBank mRNA | BC017613.1 | - | 4 | 1160 | 4912570 | |
BC094656.1 | - | 4 | 1130 |