Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC23A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005847.4 | NP_005838.3 | 14 | 1896 | ||
NM_152685.3 | NP_689898.2 | 14 | 1908 | |||
XM_005272148.3 | XP_005272205.3 | 15 | 2033 | |||
XM_005272149.3 | XP_005272206.3 | 15 | 2021 | |||
XM_006714741.2 | XP_006714804.2 | 14 | 1937 | |||
XM_011543765.2 | XP_011542067.1 | 15 | 2033 | |||
XM_011543766.1 | XP_011542068.1 | 14 | 1814 | |||
XM_011543767.1 | XP_011542069.1 | 13 | 1718 | |||
XM_011543768.1 | XP_011542070.1 | 14 | 1938 | |||
XM_011543769.1 | XP_011542071.1 | 13 | 1693 | |||
GenBank mRNA | AF098277.1 | - | 14 | 1846 | ||
AF170911.1 | - | 14 | 1830 | |||
AK092655.1 | - | 13 | 1720 | |||
AK290789.1 | - | 14 | 1829 | |||
BC019225.1 | - | 7 | 844 | |||
BC050261.1 | - | 14 | 1850 | 5188430 |