Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145421.2 | NP_663396.1 | 5 | 1223 | ||
GenBank mRNA | AK170785.1 | - | 4 | 1288 | ||
AK177751.1 | - | 1 | 136 | |||
AK220316.1 | - | 3 | 4470 | |||
BC005632.1 | - | 5 | 1198 | 3964094 | ||
BC082997.1 | - | 5 | 1187 | 4160194 |